2,4-Heptadecadiynoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0667-100MG
TCH0667-100MGEA 77.11 USD
TCH0667-100MG
2,4-Heptadecadiynoic acid ≥98.0% (by titrimetric analysis)
2,4-Heptadecadiynoic Acid

CAS Number: 64034-02-2
MDL Number: MFCD00060064
Molecular Formula: C17H26O2
Molecular Weight: 262.39
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 60
Storage Temperature: 0°C

Formula: C₁₇H₂₆O₂
Melting Pt: 60 °C
MDL Number: MFCD00060064
CAS Number: 64034-02-2
https://us.vwr.com/stibo/web/std.lang.all/06/83/28970683.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR