TCIAH0651

H0651-25G
TCH0651-025GEA 171.67 USD
TCH0651-025G
TCIAH0651
CAS Number: 4857-44-7
MDL Number: MFCD00042725
Molecular Formula: C5H10O3S
Molecular Weight: 338.45
Purity/Analysis Method: 95.0% (T)
Form: Crystal

Other Product Names / Synonyms: Calcium alpha-Hydroxy-DL-methionine, alpha-Hydroxy-DL-methionine Calcium Salt, 2-Hydroxy-4-(methylthio)butyric Acid Calcium Salt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR