2-Hexen-1-ylsuccinic anhydride (cis and trans mixture) ≥96.0% (by GC, titration analysis)
Supplier: TCI America
H0616-25G
TCH0616-025GEA
182
USD
TCH0616-025G
2-Hexen-1-ylsuccinic anhydride (cis and trans mixture) ≥96.0% (by GC, titration analysis)
2-Hexen-1-ylsuccinic anhydride (cis and trans mixture)
CAS Number: 10500-34-2
MDL Number: MFCD00047131
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 1.07
Formula:
C₁₀H₁₄O₃ MW: 182.22 g/mol Density: 1.07 (20 °C) |
MDL Number:
MFCD00047131 CAS Number: 10500-34-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...