Heptadecanenitrile ≥95.0%

Supplier: TCI America

Synonyms: HeptadecanonitrileHexadecyl cyanideCetyl cyanideHeptadecanenitrile

H0568-25G
TCH0568-025GEA 40.58 USD
TCH0568-025G
Heptadecanenitrile ≥95.0%
Heptadecanenitrile

CAS Number: 5399-02-0
MDL Number: MFCD00042635
Molecular Formula: C17H33N
Molecular Weight: 251.46
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Boiling point (°C): 208
Melting point (°C): 34


Other Product Names / Synonyms: Cetyl Cyanide, Hexadecyl Cyanide, Heptadecanenitrile

Formula: C₁₇H₃₃N
Boiling Pt: 208 °C
Melting Pt: 34 °C
MDL Number: MFCD00042635
CAS Number: 5399-02-0
EINECS: 226-432-1
https://us.vwr.com/stibo/web/std.lang.all/05/85/28970585.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR