1,5-Heptadiene (cis and trans mixture) ≥97.0%

Supplier: TCI America

H0551-5ML
TCH0551-005MLEA 224.25 USD
TCH0551-005ML
1,5-Heptadiene (cis and trans mixture) ≥97.0%
1,5-Heptadiene (cis and trans mixture)

CAS Number: 1541-23-7
MDL Number: MFCD00026998
Molecular Formula: C7H12
Molecular Weight: 96.17
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Flash Point (°C): -3
Specific Gravity (20/20): 0.72

Formula: C₇H₁₂
Density: 0,72 (20 °C)
Flash Pt: –3 °C
MDL Number: MFCD00026998
CAS Number: 1541-23-7

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR