2-Ethyl-3-hydroxy-4-pyrone

Supplier: TCI America

Synonyms: 2-Ethyl-3-hydroxy-4H-pyran-4-oneEthyl Maltol

H0550-25G
TCH0550-025GEA 29.44 USD
TCH0550-025G
2-Ethyl-3-hydroxy-4-pyrone
2-Ethyl-3-hydroxy-4-pyrone

CAS Number: 4940-11-8
MDL Number: MFCD00059795
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Form: Crystal
Color: White
Melting point (°C): 92


Other Product Names / Synonyms: 2-Ethyl-3-hydroxy-4H-pyran-4-one, Ethyl Maltol

Formula: C₇H₈O₃
MW: 140.14 g/mol
Melting Pt: 92 °C
MDL Number: MFCD00059795
CAS Number: 4940-11-8
https://us.vwr.com/stibo/web/std.lang.all/05/61/28970561.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR