Ethyl palmitoleate ≥95.0%
Supplier: TCI America
Synonyms:
Palmitoleic acid ethyl ester
, Ethyl cis-9-Hexadecenoate
, cis-9-Hexadecenoic acid ethyl ester
, Ethyl Palmitoleate
, (Z)-Ethyl hexadec-9-enoate
CAS Number: 56219-10-4
MDL Number: MFCD00056314
Molecular Formula: C18H34O2
Molecular Weight: 282.47
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 135
Specific Gravity (20/20): 0.87
Storage Temperature:
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Ethyl Palmitoleate, cis-9-Hexadecenoic Acid Ethyl Ester, Palmitoleic Acid Ethyl Ester
Formula:
C₁₈H₃₄O₂ MW: 282.47 g/mol Boiling Pt: 135 °C Density: 0.87 (20 °C) |
MDL Number:
MFCD00056314 CAS Number: 56219-10-4 EINECS: 260-065-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...