11-ɑ-Hydroxyprogesterone acetate ≥98.0%

Synonyms: 11alpha-Acetoxypregn-4-ene-3,20-dione11alpha-Acetoxyprogesterone11alpha-Hydroxypregn-4-ene-3,20-dione Acetate

H0499-1G
TCH0499-001GEA 100.94 USD
TCH0499-001G
11-ɑ-Hydroxyprogesterone acetate ≥98.0%
11-alpha-Hydroxyprogesterone acetate

CAS Number: 2268-98-6
MDL Number: MFCD00021152
Molecular Formula: C23H32O4
Molecular Weight: 372.51
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 177


Product Categories: Bioscience, Pregnanes (Progestogen), Steroids
Other Product Names / Synonyms: 11alpha-Acetoxypregn-4-ene-3,20-dione, 11alpha-Acetoxyprogesterone, 11alpha-Hydroxypregn-4-ene-3,20-dione Acetate

Formula: C₂₃H₃₂O₄
MW: 372.51 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00021152
CAS Number: 2268-98-6
https://us.vwr.com/stibo/web/std.lang.all/05/09/28970509.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR