1-(2-Hydroxyethyl)pyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-(2-Hydroxyethyl)pyrrolidine
, 1-Pyrrolidineethanol
H0479-25ML
TCH0479-025MLEA
53.78
USD
TCH0479-025ML
1-(2-Hydroxyethyl)pyrrolidine ≥98.0% (by GC, titration analysis)
1-(2-Hydroxyethyl)pyrrolidine
CAS Number: 2955-88-6
MDL Number: MFCD00003181
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 187
Flash Point (°C): 56
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 1-Pyrrolidineethanol
Formula:
C₆H₁₃NO MW: 115.18 g/mol Boiling Pt: 187 °C Density: 0.98 (20 °C) Flash Pt: 56 °C |
MDL Number:
MFCD00003181 CAS Number: 2955-88-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...