(-)-Hyoscyamine ≥99.0% (by titrimetric analysis)

Synonyms: L-Hyoscyamine

H0450-5G
TCH0450-005GEA 131.58 USD
TCH0450-005G
(-)-Hyoscyamine ≥99.0% (by titrimetric analysis)
(-)-Hyoscyamine

CAS Number: 101-31-5
MDL Number: MFCD00067306
Molecular Formula: C17H23NO3
Molecular Weight: 289.38
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 108
Specific rotation [a]20/D: -22 deg (C=1, EtOH(50vol%))

Formula: C₁₇H₂₃NO₃
Melting Pt: 108 °C
MDL Number: MFCD00067306
CAS Number: 101-31-5
https://us.vwr.com/stibo/web/std.lang.all/04/66/28970466.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR