(2-Hydroxyethyl)triethylammonium Iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Triethylcholine IodideN,N,N-Triethyl-2-hydroxyethanaminium iodide

H0407-10G
TCH0407-010GEA 114.65 USD
TCH0407-010G
(2-Hydroxyethyl)triethylammonium Iodide ≥98.0% (by titrimetric analysis)
(2-Hydroxyethyl)triethylammonium Iodide

CAS Number: 5957-17-5
MDL Number: MFCD00031691
Molecular Formula: C8H20INO
Molecular Weight: 273.16
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Triethylcholine Iodide

Formula: C₈H₂₀INO
MW: 273.16 g/mol
MDL Number: MFCD00031691
CAS Number: 5957-17-5
https://us.vwr.com/stibo/web/std.lang.all/04/26/28970426.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR