7-(2-Hydroxyethyl)theophylline ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
H0402-500G
H0402-25G
TCH0402-025GEA
89.29
USD
TCH0402-025G
TCH0402-500G
7-(2-Hydroxyethyl)theophylline ≥98.0% (by HPLC, total nitrogen)
7-(2-Hydroxyethyl)theophylline
CAS Number: 519-37-9
MDL Number: MFCD00055055
Molecular Formula: C9H12N4O3
Molecular Weight: 224.22
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 163
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients, Nucleobases and Their Analogs, Purine Alkaloids
Other Product Names / Synonyms: Etofylline
Formula:
C₉H₁₂N₄O₃ Melting Pt: 163 °C |
MDL Number:
MFCD00055055 CAS Number: 519-37-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...