1-(2-Hydroxyethyl)-4-(3-hydroxypropyl)piperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Ethanol-4-propanolpiperidine

H0360-25G
TCH0360-025GEA 148.14 USD
TCH0360-025G
1-(2-Hydroxyethyl)-4-(3-hydroxypropyl)piperidine ≥98.0% (by GC, titration analysis)
1-(2-Hydroxyethyl)-4-(3-hydroxypropyl)piperidine

CAS Number: 19780-85-9
MDL Number: MFCD00047481
Molecular Formula: C10H21NO2
Molecular Weight: 187.28
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 49


Other Product Names / Synonyms: 1-Ethanol-4-propanolpiperidine

Formula: C₁₀H₂₁NO₂
MW: 187.28 g/mol
Melting Pt: 49 °C
MDL Number: MFCD00047481
CAS Number: 19780-85-9
https://us.vwr.com/stibo/web/std.lang.all/03/94/28970394.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR