(±)-δ-Undecalactone ≥98.0%

Supplier: TCI America

Synonyms: (±)-Tetrahydro-6-hexyl-2H-pyran-2-oneUndecanoic δ-lactonedelta-Undecanolactone(±)-Undecan-5-olide6-Hexyltetrahydro-2H-pyran-2-one5-Hydroxyundecanoic acid delta-Lactone

H0344-25G
TCH0344-025GEA 40.58 USD
TCH0344-025G
(±)-δ-Undecalactone ≥98.0%
(±)-δ-Undecalactone

CAS Number: 710-04-3
MDL Number: MFCD00006650
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Flash Point (°C): 110
Specific Gravity (20/20): 0.96


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 5-Hydroxyundecanoic Acid delta-Lactone, delta-Undecalactone

Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 168 °C
Density: 0.96 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00006650
CAS Number: 710-04-3
EINECS: 211-915-1
https://us.vwr.com/stibo/web/std.lang.all/03/83/28970383.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR