8-Hydroxy-7-iodoquinoline-5-sulfonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Ferron7-Iodo-8-hydroxyquinoline-5-sulfonic acid7-Iodo-8-hydroxyquinoline-5-sulphonic acidLoretin

H0254-25G
TCH0254-025GEA 204.09 USD
TCH0254-025G
8-Hydroxy-7-iodoquinoline-5-sulfonic acid ≥98.0% (by HPLC, titration analysis)
8-Hydroxy-7-iodoquinoline-5-sulfonic acid

CAS Number: 547-91-1
MDL Number: MFCD00006793
Molecular Formula: C9H6INO4S
Molecular Weight: 351.11
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 270


Other Product Names / Synonyms: Ferron, 7-Iodo-8-hydroxyquinoline-5-sulfonic Acid, Loretin

Formula: C₉H₆INO₄S
MW: 351.11 g/mol
Melting Pt: 270 °C
MDL Number: MFCD00006793
CAS Number: 547-91-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR