DL-2-Hydroxybutyric acid ≥95.0% (by titrimetric analysis) (contains Polymolecular esterification product)

Supplier: TCI America

Synonyms: 2-Hydroxybutanoic acid

H0227-25G
TCH0227-025GEA 153.21 USD
TCH0227-025G
DL-2-Hydroxybutyric acid ≥95.0% (by titrimetric analysis) (contains Polymolecular esterification product)
DL-2-Hydroxybutyric acid

CAS Number: 600-15-7
MDL Number: MFCD00070502
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Boiling point (°C): 260
Melting point (°C): 43
Storage Temperature: 0-10°C

Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 260 °C
Melting Pt: 43 °C
MDL Number: MFCD00070502
CAS Number: 600-15-7
https://us.vwr.com/stibo/web/std.lang.all/02/71/28970271.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR