Phenyl-4-hydroxybenzoate ≥99.0%

Supplier: TCI America

H0218-25G
TCH0218-025GEA 93.38 USD
TCH0218-025G
Phenyl-4-hydroxybenzoate ≥99.0%
Phenyl 4-hydroxybenzoate

CAS Number: 17696-62-7
MDL Number: MFCD00016467
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 182


Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Phenyl Ester, Phenylparaben

Formula: C₁₃H₁₀O₃
Melting Pt: 182 °C
MDL Number: MFCD00016467
CAS Number: 17696-62-7
https://us.vwr.com/stibo/web/std.lang.all/02/63/28970263.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR