Glycolic acid nitrile ∼52% ∼9.7 mol/L in water stabilized

Supplier: TCI America

Synonyms: Formaldehyde CyanhydrinHydroxyacetonitrileGlycolonitrile

H0191-25G
TCH0191-025GEA 157 USD
TCH0191-025G
Glycolic acid nitrile ∼52% ∼9.7 mol/L in water stabilized
Glycolic acid nitrile

(stabilized with H2SO4)
CAS Number: 107-16-4
MDL Number: MFCD00042731
Molecular Formula: C2H3NO
Molecular Weight: 57.05
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 183
Melting point (°C): -67
Flash Point (°C): 69
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Formaldehyde Cyanhydrin, Hydroxyacetonitrile

https://us.vwr.com/stibo/web/std.lang.all/02/38/28970238.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR