Di-n-alkyl adipate

Synonyms: Adipic Acid Di-n-alkyl Ester

A1386-25ML
TCA1386-025MLEA 30.9 USD
TCA1386-025ML
Di-n-alkyl adipate
Di-n-alkyl adipate

[Plasticizer]
MDL Number: MFCD00675047
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: Adipic Acid Di-n-alkyl Ester

https://us.vwr.com/stibo/web/std.lang.all/93/16/28929316.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR