Isopentyl hexanoate ≥97.0% (contains 2-Methylbutyl Hexanoate)

Supplier: TCI America

Synonyms: Isopentyl hexanoateHexanoic acid isoamyl esterIsoamyl caproateCaproic acid isoamyl esterCaproic acid isopentyl esterIsoamyl hexanoate3-Methylbutyl hexanoateHexanoic acid isopentyl esterHexanoic acid Isopentyl esterIsopentyl caproate

H0109-25ML
TCH0109-025MLEA 27.41 USD
TCH0109-025ML
Isopentyl hexanoate ≥97.0% (contains 2-Methylbutyl Hexanoate)
Isopentyl hexanoate

CAS Number: 2198-61-0
MDL Number: MFCD00027280
Molecular Formula: C11H22O2
Molecular Weight: 186.30
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 88
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Hexanoic Acid Isopentyl Ester, Isopentyl Hexanoate, Hexanoic Acid Isoamyl Ester

Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
Boiling Pt: 98 °C
Density: 0.86 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00027280
CAS Number: 2198-61-0
EINECS: 218-600-8
https://us.vwr.com/stibo/web/std.lang.all/01/71/28970171.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR