TCIAA1382

A1382-1G
TCA1382-001GEA 185.28 USD
TCA1382-001G
TCIAA1382
CAS Number: 7250-68-2
MDL Number: MFCD00096305
Molecular Formula: C18H18N2O4
Molecular Weight: 326.35
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 144

Other Product Names / Synonyms: 4,4'-Azodibenzoic Acid Diethyl Ester, Diethyl Azobenzene-4,4'-dicarboxylate, Azobenzene-4,4'-dicarboxylic Acid Diethyl Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR