alpha,alpha',2,4,5,6-Hexachloro-m-xylene ≥98.0%

H0063-25G
TCH0063-025GEA 157.08 USD
TCH0063-025G
alpha,alpha',2,4,5,6-Hexachloro-m-xylene ≥98.0%
alpha,alpha',2,4,5,6-Hexachloro-m-xylene

CAS Number: 1133-57-9
MDL Number: MFCD00039355
Molecular Formula: C8H4Cl6
Molecular Weight: 312.82
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 139

Formula: C₈H₄Cl₆
MW: 312.82 g/mol
Melting Pt: 139 °C
MDL Number: MFCD00039355
CAS Number: 1133-57-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR