Galβ(1-3)GalNAc-β-pNP ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-Nitrophenyl 2-Acetamido-2-deoxy-3-O-(β-D-galactopyranosyl)-β-D-galactopyranoside
CAS Number: 59837-15-9
Molecular Formula: C20H28N2O13
Molecular Weight: 504.45
Purity/Analysis Method:
Form: Crystal
Storage Temperature:
Product Categories: Synthetic Organic Chemistry, p-Nitrophenyl Glycosides, Bioscience, Disaccharides, Glycosides, p-Nitrophenyl Glycosides
Other Product Names / Synonyms: 4-Nitrophenyl 2-Acetamido-2-deoxy-3-O-(beta-D-galactopyranosyl)-beta-D-galactopyranoside
Formula:
C₂₀H₂₈N₂O₁₃ MW: 504.45 g/mol |
CAS Number:
59837-15-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...