5-Methyl-3-heptyl acetate ≥95.0%

Synonyms: 1-Ethyl-3-methylpentyl AcetateAcetic Acid 1-Ethyl-3-methylpentyl EsterAcetic Acid 5-Methyl-3-heptyl Ester

A1379-5G
TCA1379-005GEA 130.56 USD
TCA1379-005G
5-Methyl-3-heptyl acetate ≥95.0%
5-Methyl-3-heptyl acetate

CAS Number: 375855-08-6
MDL Number: MFCD00671490
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Acetic Acid 1-Ethyl-3-methylpentyl Ester, Acetic Acid 5-Methyl-3-heptyl Ester, 1-Ethyl-3-methylpentyl Acetate

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 185 °C
Density: 0.87 (20 °C)
MDL Number: MFCD00671490
CAS Number: 375855-08-6
https://us.vwr.com/stibo/web/std.lang.all/93/07/28929307.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR