Dipotassium glycyrrhizinate hydrate ≥75.0% (by HPLC)

Supplier: TCI America

Synonyms: Glycyrrhizic Acid Dipotassium SaltGlycyrrhizin Dipotassium Salt

G0270-25G
TCG0270-025GEA 118.35 USD
TCG0270-025G
Dipotassium glycyrrhizinate hydrate ≥75.0% (by HPLC)
Dipotassium glycyrrhizinate hydrate

CAS Number: 68797-35-3
MDL Number: MFCD02094742
Molecular Formula: C42H62O16
Molecular Weight: 899.12
Purity/Analysis Method: 75.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 52 deg (C=1.5, H2O)


Product Categories: Bioscience, Obesity/Diabete Research Reagents, Glycosides, Disaccharides, Glycosides, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Triterpenes
Other Product Names / Synonyms: Glycyrrhizic Acid Dipotassium Salt, Glycyrrhizin Dipotassium Salt

Formula: C₄₂H₆₂O₁₆
MW: 899.12 g/mol
MDL Number: MFCD02094742
CAS Number: 68797-35-3
https://us.vwr.com/stibo/web/std.lang.all/98/98/28969898.jpg

Order Now

Specification Test Results

Purity/Analysis Method 75.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR