Acid green 50
Supplier: TCI America
Synonyms:
Green S
, Lissamine Green B
G0250-25G
TCG0250-25GEA
45.68
USD
TCG0250-25G
Acid green 50
Acid green 50
CAS Number: 3087-16-9
MDL Number: MFCD00012117
Molecular Formula: C27H26N2O7S2
Molecular Weight: 576.61
Form: Crystal
Color: Deep Yellow Red
Other Product Names / Synonyms: Green S, Lissamine Green B
Formula:
C₂₇H₂₆N₂O₇S₂ MW: 576.61 g/mol |
MDL Number:
MFCD00012117 CAS Number: 3087-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...