Glycine-N,N-bis(methylenephosphonic acid) ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N,N-Bis(phosphonomethyl)glycine

G0229-5G
TCG0229-005GEA 71.03 USD
TCG0229-005G
Glycine-N,N-bis(methylenephosphonic acid) ≥97.0% (by titrimetric analysis)
Glycine-N,N-bis(methylenephosphonic acid)

CAS Number: 2439-99-8
MDL Number: MFCD00056670
Molecular Formula: C4H11NO8P2
Molecular Weight: 263.08
Purity/Analysis Method: 97.0% (T)
Form: Crystal


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: N,N-Bis(phosphonomethyl)glycine

Formula: C₄H₁₁NO₈P₂
MW: 263.08 g/mol
MDL Number: MFCD00056670
CAS Number: 2439-99-8
https://us.vwr.com/stibo/web/std.lang.all/98/66/28969866.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR