D-Glucuronamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

G0223-25G
TCG0223-025GEA 182.35 USD
TCG0223-025G
D-Glucuronamide ≥98.0% (by total nitrogen basis)
D-Glucuronamide

CAS Number: 3789-97-7
MDL Number: MFCD00006619
Molecular Formula: C6H11NO6
Molecular Weight: 193.16
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 167
Specific rotation [a]20/D: 33 deg (C=2, H2O)

Formula: C₆H₁₁NO₆
Melting Pt: 167 °C
MDL Number: MFCD00006619
CAS Number: 3789-97-7
https://us.vwr.com/stibo/web/std.lang.all/98/62/28969862.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR