Geranyllinalool (mixture of isomers) ≥90.0%
Supplier: TCI America
Synonyms:
3,7,11,15-Tetramethyl-1,6,10,14-hexadecatetraen-3-ol
CAS Number: 68931-30-6
MDL Number: MFCD00059363
Molecular Formula: C20H34O
Molecular Weight: 290.49
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 110
Specific Gravity (20/20): 0.88
Product Categories: Bioscience, Diterpenes
Other Product Names / Synonyms: 3,7,11,15-Tetramethyl-1,6,10,14-hexadecatetraen-3-ol
Formula:
C₂₀H₃₄O MW: 290.49 g/mol Boiling Pt: 140 °C Density: 0.88 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00059363 CAS Number: 68931-30-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...