DL-Alanyl-DL-valine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
H-DL-Ala-DL-Val-OH
A0192-100MG
TCA0192-100MGEA
85
USD
TCA0192-100MG
DL-Alanyl-DL-valine ≥98.0% (by titrimetric analysis)
DL-Alanyl-DL-valine
CAS Number: 1999-46-8
MDL Number: MFCD00038163
Molecular Formula: C8H16N2O3
Molecular Weight: 188.23
Purity/Analysis Method:
Form: Crystal
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-DL-Ala-DL-Val-OH
Formula:
C₈H₁₆N₂O₃ MW: 188.23 g/mol |
MDL Number:
MFCD00038163 CAS Number: 1999-46-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...