Calcium glycolate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Glycolic Acid Calcium SaltHydroxyacetic Acid Calcium Salt

G0200-25G
TCG0200-025GEA 79 USD
TCG0200-025G
Calcium glycolate ≥97.0% (by titrimetric analysis)
Calcium glycolate

CAS Number: 996-23-6
MDL Number: MFCD00067202
Molecular Formula: C2H4O3
Molecular Weight: 190.16
Purity/Analysis Method: 97.0% (T)
Form: Crystal


Other Product Names / Synonyms: Glycolic Acid Calcium Salt, Hydroxyacetic Acid Calcium Salt

Formula: C₂H₄O₃
MW: 190.16 g/mol
MDL Number: MFCD00067202
CAS Number: 996-23-6
https://us.vwr.com/stibo/web/std.lang.all/98/45/28969845.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR