Butyl gallate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Gallic Acid Butyl Ester
G0186-25G
TCG0186-025GEA
100.45
USD
TCG0186-025G
Butyl gallate ≥98.0% (by GC, titration analysis)
Butyl gallate
CAS Number: 1083-41-6
MDL Number: MFCD00016432
Molecular Formula: C11H14O5
Molecular Weight: 226.23
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 144
Other Product Names / Synonyms: Gallic Acid Butyl Ester
Formula:
C₁₁H₁₄O₅ MW: 226.23 g/mol Melting Pt: 144 °C |
MDL Number:
MFCD00016432 CAS Number: 1083-41-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...