Glycyl-DL-serine hydrate
Synonyms:
H-Gly-DL-Ser-OH
G0182-1G
TCG0182-001GEA
192.03
USD
TCG0182-001G
Glycyl-DL-serine hydrate
Glycyl-DL-serine hydrate
CAS Number: 687-38-7
MDL Number: MFCD00025583
Molecular Formula: C5H10N2O4
Molecular Weight: 162.15
Form: Crystal
Color: White
Other Product Names / Synonyms: H-Gly-DL-Ser-OH
Formula:
C₅H₁₀N₂O₄ MW: 162.15 g/mol |
MDL Number:
MFCD00025583 CAS Number: 687-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...