Glyoxal bis(sodium hydrogen sulfite) monohydrate ≥97.0% (by titrimetric analysis) (contains oligomers)
Supplier: TCI America
Synonyms:
Glyoxal sodium Bisulfite hydrate
, Glyoxal sodium Bisulphite hydrate
G0154-25G
G0154-500G
TCG0154-025GEA
16.24
USD
TCG0154-025G
TCG0154-500G
Glyoxal bis(sodium hydrogen sulfite) monohydrate ≥97.0% (by titrimetric analysis) (contains oligomers)
Glyoxal bis(sodium hydrogen sulphite) monohydrate
CAS Number: 517-21-5
MDL Number: MFCD00007516
Molecular Formula: C2H2O2
Molecular Weight: 266.15
Purity/Analysis Method:
Form: Crystal
Formula:
C₂H₄Na₂O₈S₂ MW: 266.16 g/mol |
MDL Number:
MFCD00007516 CAS Number: 517-21-5 EINECS: 208-233-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...