Monoammonium glycyrrhizinate hydrate ≥75.0% (by HPLC)
Supplier: TCI America
Synonyms:
Glycyrrhizic Acid Monoammonium Salt
CAS Number: 53956-04-0
MDL Number: MFCD00167400
Molecular Formula: C42H65NO16
Molecular Weight: 839.97
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 217
Specific rotation [a]20/D: 49 deg (C=1.5, EtOH)
Product Categories: Bioscience, Obesity/Diabete Research Reagents, Glycosides, Disaccharides, Glycosides, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Triterpenes
Other Product Names / Synonyms: Glycyrrhizic Acid Monoammonium Salt, Glycyrrhizin Monoammonium Salt
Formula:
C₄₂H₆₅NO₁₆ MW: 839.97 g/mol (anhy) Melting Pt: 217 °C |
MDL Number:
MFCD00167400 CAS Number: 53956-04-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...