Glycyrrhizin ≥93.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Glycyrrhizic Acid

G0150-1G G0150-25G
TCG0150-025GEA 277.02 USD
TCG0150-025G TCG0150-001G
Glycyrrhizin ≥93.0% (by titrimetric analysis)
Glycyrrhizin

CAS Number: 1405-86-3
MDL Number: MFCD00065194
Molecular Formula: C42H62O16
Molecular Weight: 822.94
Purity/Analysis Method: 93.0% (T)
Form: Crystal
Specific rotation [a]20/D: 61 deg (C=1.5, EtOH)


Product Categories: Bioscience, Obesity/Diabete Research Reagents, Glycosides, Disaccharides, Glycosides, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Triterpenes
Other Product Names / Synonyms: Glycyrrhizic Acid

Formula: C₄₂H₆₂O₁₆
MW: 822.94 g/mol
MDL Number: MFCD00065194
CAS Number: 1405-86-3
https://us.vwr.com/stibo/web/std.lang.all/98/11/28969811.jpg

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR