Glycylsarcosine ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
H-Gly-Sar-OH
G0138-1G
G0138-5G
TCG0138-5GEA
411.08
USD
TCG0138-5G
TCG0138-001G
Glycylsarcosine ≥97.0% (by HPLC)
Glycylsarcosine
CAS Number: 29816-01-1
MDL Number: MFCD00038193
Molecular Formula: C5H10N2O3
Molecular Weight: 146.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 202
Storage Temperature: 0-10°C
Formula:
C₅H₁₀N₂O₃ MW: 146.15 g/mol Melting Pt: 202 °C |
MDL Number:
MFCD00038193 CAS Number: 29816-01-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...