Glycyl-L-Isoleucine

Supplier: TCI
G0129-1G
TCG0129-001GEA 118.32 USD
TCG0129-001G
Glycyl-L-Isoleucine
Proteins and Peptides
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Ile-OH

  • Melting point: 250 °C
  • Specific rotation [a]20/D: –14° (C=2, H₂O)
  • EINECS: 243-085-1
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR