1,3-Adamantanediacetic acid ≥97.0%
Supplier: TCI America
Synonyms:
1,3-Bis(carboxymethyl)adamantane
A1357-5G
TCA1357-005GEA
179.66
USD
TCA1357-005G
1,3-Adamantanediacetic acid ≥97.0%
1,3-Adamantanediacetic acid
CAS Number: 17768-28-4
MDL Number: MFCD00191708
Molecular Formula: C14H20O4
Molecular Weight: 252.31
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 237
Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 1,3-Bis(carboxymethyl)adamantane
Formula:
C₁₄H₂₀O₄ MW: 252.31 g/mol Melting Pt: 237 °C |
MDL Number:
MFCD00191708 CAS Number: 17768-28-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...