1-(3-Acetamidophenyl)-5-mercaptotetrazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-(3-(5-Mercapto-1H-tetrazol-1-yl)phenyl)acetamide

A1352-10G
TCA1352-010GEA 101.47 USD
TCA1352-010G
1-(3-Acetamidophenyl)-5-mercaptotetrazole ≥98.0% (by titrimetric analysis)
1-(3-Acetamidophenyl)-5-mercaptotetrazole

CAS Number: 14070-48-5
MDL Number: MFCD00603728
Molecular Formula: C9H9N5OS
Molecular Weight: 235.27
Purity/Analysis Method: 98.0% (T)
Form: Crystal

Formula: C₉H₉N₅OS
MW: 235.27 g/mol
MDL Number: MFCD00603728
CAS Number: 14070-48-5
https://us.vwr.com/stibo/web/std.lang.all/92/69/28929269.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR