Girard's reagent P (carbazoylmethylpyridinium chloride) ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(1-Pyridinio)acetohydrazide chloride
, 1-(2-Hydrazinyl-2-oxoethyl)pyridin-1-ium chloride
, 1-(Hydrazinocarbonylmethyl)pyridinium chloride
, Girard's Reagent P
CAS Number: 1126-58-5
MDL Number: MFCD00011992
Molecular Formula: C7H10ClN3O
Molecular Weight: 187.63
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 203
Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: 1-(Hydrazinocarbonylmethyl)pyridinium Chloride, (1-Pyridinio)acetohydrazide Chloride
Formula:
C₇H₁₀ClN₃O MW: 187.63 g/mol Melting Pt: 203 °C |
MDL Number:
MFCD00011992 CAS Number: 1126-58-5 EINECS: 214-421-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...