Gentiobiose ≥96.0% (by HPLC)

Supplier: TCI America

Synonyms: β-D-Gentiobioseβ-Gentiobiose

G0026-100MG
TCG0026-100MGEA 130.63 USD
TCG0026-100MG
Gentiobiose ≥96.0% (by HPLC)
Gentiobiose

CAS Number: 554-91-6
MDL Number: MFCD00198056
Molecular Formula: C12H22O11
Molecular Weight: 342.30
Purity/Analysis Method: 96.0% (HPLC)
Form: Crystal
Melting point (°C): 197
Specific rotation [a]20/D: 9.5 deg (C=4, H2O)

Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Melting Pt: 197 °C
MDL Number: MFCD00198056
CAS Number: 554-91-6
https://us.vwr.com/stibo/web/std.lang.all/97/27/28969727.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR