Fluorotrimethylsilylketene ethyl trimethylsilyl acetal (mixture of isomers) ≥95.0%

F0692-5G
TCF0692-5GEA 465.12 USD
TCF0692-5G
Fluorotrimethylsilylketene ethyl trimethylsilyl acetal (mixture of isomers) ≥95.0%
Fluorotrimethylsilylketene ethyl trimethylsilyl acetal (mixture of isomers)

CAS Number: 1068142-02-8
Molecular Formula: C10H23FO2Si2
Molecular Weight: 250.46
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C

Formula: C₁₀H₂₃FO₂Si₂
Density: 0.92 (20 °C)
CAS Number: 1068142-02-8
https://us.vwr.com/stibo/web/std.lang.all/92/30/28969230.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR