1,1'-Ferrocenediboronic acid ≥95.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
1,1'-Diborono-Ferrocene
F0664-1G
TCF0664-1GEA
141.04
USD
TCF0664-1G
1,1'-Ferrocenediboronic acid ≥95.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
1,1'-Ferrocenediboronic acid
CAS Number: 32841-83-1
Molecular Formula: C10H12B2FeO4
Molecular Weight: 273.67
Purity/Analysis Method:
Form: Crystal
Formula:
C₁₀H₁₂B₂FeO₄ MW: 273.67 g/mol |
MDL Number:
MFCD00799542 CAS Number: 32841-83-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...