Furfuryl methyl disulfide ≥98.0%
Supplier: TCI America
Synonyms:
Methyl furfuryl disulphide
, 2-((Methyldisulphanyl)methyl)furan
, Methyl furfuryl disulfide
, 2-((Methyldisulfanyl)methyl)furan
F0561-5G
F0561-25G
TCF0561-25GEA
113.22
USD
TCF0561-25G
TCF0561-5G
Furfuryl methyl disulfide ≥98.0%
Furfuryl methyl disulphide
CAS Number: 57500-00-2
MDL Number: MFCD00012326
Molecular Formula: C6H8OS2
Molecular Weight: 160.25
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 61
Flash Point (°C): 90
Specific Gravity (20/20): 1.19
Formula:
C₆H₈OS₂ MW: 160.2571 g/mol Boiling Pt: 61 °C Density: 1.19 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00012326 CAS Number: 57500-00-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...