5'-Formyl-2,2'-bithiophene-5-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 5-(5-Formyl-2-thienyl)-2-thiopheneboronic Acid

F0548-1G
TCF0548-1GEA 471.76 USD
TCF0548-1G
5'-Formyl-2,2'-bithiophene-5-boronic acid (contains varying amounts of Anhydride)
5'-Formyl-2,2'-bithiophene-5-boronic acid

CAS Number: 1369328-62-0
Molecular Formula: C9H7BO3S2
Molecular Weight: 238.08
Form: Crystal
Color: Pale Yellow
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 5-(5-Formyl-2-thienyl)-2-thiopheneboronic Acid

Formula: C₉H₇BO₃S₂
MW: 238.08 g/mol
CAS Number: 1369328-62-0
https://us.vwr.com/stibo/web/std.lang.all/91/12/28969112.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR