4'-Formylbenzo-18-crown-6 (4'-formylbenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥98.0%

Supplier: TCI America

Synonyms: 2,3-(4-Formylbenzo)-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene4'-Formylbenzo-18-crown 6-Ether

F0451-500MG
TCF0451-500MGEA 1233 USD
TCF0451-500MG
4'-Formylbenzo-18-crown-6 (4'-formylbenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥98.0%
4'-Formylbenzo-18-crown-6 (4'-Formylbenzo-1,4,7,10,13,16-hexoxacyclooctadecane)

CAS Number: 60835-74-7
MDL Number: MFCD00388882
Molecular Formula: C17H24O7
Molecular Weight: 340.37
Purity/Analysis Method: 98.0% (GC)
Form: Crystal


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-(4-Formylbenzo)-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene

https://us.vwr.com/stibo/web/std.lang.all/90/40/28969040.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR