Ethyl-2-formylpropionate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Formylpropionic Acid Ethyl Ester

F0411-1G F0411-5G
TCF0411-5GEA 814.8 USD
TCF0411-5G TCF0411-001G
Ethyl-2-formylpropionate ≥97.0% (by titrimetric analysis)
Ethyl-2-formylpropionate

CAS Number: 27772-62-9
MDL Number: MFCD00044012
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: 97.0% (T)
Form: Clear Liquid
Storage Temperature: 0°C


Other Product Names / Synonyms: 2-Formylpropionic Acid Ethyl Ester

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD00044012
CAS Number: 27772-62-9
https://us.vwr.com/stibo/web/std.lang.all/90/06/28969006.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR