5-(4-Fluorophenoxy)valeric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0410-5G
TCF0410-005GEA 335 USD
TCF0410-005G
5-(4-Fluorophenoxy)valeric acid ≥98.0% (by GC, titration analysis)
5-(4-Fluorophenoxy)valeric acid

CAS Number: 347867-75-8
MDL Number: MFCD02093469
Molecular Formula: C11H13FO3
Molecular Weight: 212.22
Purity/Analysis Method: 98.0% (T,GC)
Form: Crystal
Melting point (°C): 80

Formula: C₁₁H₁₃FO₃
Melting Pt: 80 °C
MDL Number: MFCD02093469
CAS Number: 347867-75-8
https://us.vwr.com/stibo/web/std.lang.all/90/05/28969005.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR