TCIAF0377

F0377-5ML
TCF0377-005MLEA 161.41 USD
TCF0377-005ML
TCIAF0377
CAS Number: 144977-47-9
MDL Number: MFCD01631308
Molecular Formula: C17H23F
Molecular Weight: 246.37
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.98
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR